Assay Design



Gene Transcripts

Gene Symbol
ntl-3
Entrez Gene ID
176614
Gene Name
protein-coding
Gene Aliases
CELE_Y56A3A.1
Chromosome Location
Chr.III: 11830907 - 11847353 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083183.3 NP_001076652.1 3 - 4 725 82

View Full Product Details