Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001268248.4 | NP_001255177.1 | 9 - 10 | 1860 | 82 |
NM_001268249.2 | NP_001255178.1 | 9 - 10 | 1867 | 82 | |
NM_001312958.3 | NP_001299887.1 | 9 - 10 | 1876 | 82 | |
NM_001312959.3 | NP_001299888.1 | 9 - 10 | 1869 | 82 | |
NM_001312960.3 | NP_001299889.1 | 6 - 7 | 1290 | 82 | |
NM_001312961.3 | NP_001299890.1 | 6 - 7 | 1290 | 82 | |
NM_001312962.3 | NP_001299891.1 | 6 - 7 | 1281 | 82 | |
NM_001312963.3 | NP_001299892.1 | 6 - 7 | 1281 | 82 | |
NM_001312964.3 | NP_001299893.1 | 5 - 6 | 720 | 82 | |
NM_001312965.3 | NP_001299894.1 | 5 - 6 | 720 | 82 | |
NM_001312966.3 | NP_001299895.1 | 5 - 6 | 711 | 82 | |
NM_001312967.3 | NP_001299896.1 | 5 - 6 | 711 | 82 | |
NM_001372667.1 | NP_001360800.1 | 9 - 10 | 1876 | 82 |