Assay Design
Gene Transcripts
- Gene Symbol
- elpc-2
- Entrez Gene ID
-
176687
- Gene Name
- ELongator complex Protein Component
- Gene Aliases
-
CELE_Y111B2A.17
- Chromosome Location
-
Chr.III: 12669043 - 12686248 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067247.10 |
NP_499648.3 |
9 - 10 |
2116 |
66 |
View Full Product Details