Assay Design
Gene Transcripts
- Gene Symbol
- lis-1
- Entrez Gene ID
-
176758
- Gene Name
- Lissencephaly-1 homolog
- Gene Aliases
-
CELE_T03F6.5
- Chromosome Location
-
Chr.III: 13370922 - 13376710 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067354.7 |
NP_499755.1 |
9 - 10 |
1214 |
62 |
GenBank mRNA |
AF164430.1 |
- |
9 - 10 |
1205 |
62 |
View Full Product Details