Assay Design
Gene Transcripts
- Gene Symbol
- trf-1
- Entrez Gene ID
-
176767
- Gene Name
- TNF Receptor Associated Factor (TRAF) homolog
- Gene Aliases
-
CELE_F45G2.6
- Chromosome Location
-
Chr.III: 13430462 - 13433438 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067372.5 |
NP_499773.2 |
6 - 7 |
828 |
93 |
GenBank mRNA |
AF348168.1 |
- |
6 - 7 |
753 |
93 |
View Full Product Details