Assay Design
Gene Transcripts
- Gene Symbol
- mdt-29
- Entrez Gene ID
-
176817
- Gene Name
- Mediator of RNA polymerase II transcription subunit 29
- Gene Aliases
-
CELE_K08E3.8
- Chromosome Location
-
Chr.III: 13771508 - 13778908 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067446.7 |
NP_499847.1 |
8 - 9 |
1279 |
170 |
- Gene Symbol
- pot-3
- Entrez Gene ID
-
181793
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_3R5.1
- Chromosome Location
-
Chr.III: 13773449 - 13781133 on Build WBcel235
View Full Product Details