Assay Design



Gene Transcripts

Gene Symbol
frm-5.1
Entrez Gene ID
176822
Gene Name
protein-coding
Gene Aliases
CELE_Y38C1AB.8
Chromosome Location
Chr.IV: 32463 - 48880 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268287.2 NP_001255216.1 2 - 3 217 103
NM_001268288.4 NP_001255217.2 2 - 3 217 103

View Full Product Details