Assay Design



Gene Transcripts

Gene Symbol
daf-1
Entrez Gene ID
176829
Gene Name
protein-coding
Gene Aliases
CELE_F29C4.1
Chromosome Location
Chr.IV: 132416 - 138084 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027988.5 NP_001023159.1 7 - 8 1703 81
NM_001027989.5 NP_001023160.1 7 - 8 1697 81

View Full Product Details