Assay Design
Gene Transcripts
- Gene Symbol
- dsc-4
- Entrez Gene ID
-
176852
- Gene Name
- Defecation Suppressor of Clk-1
- Gene Aliases
-
CELE_K02D7.4
- Chromosome Location
-
Chr.IV: 315056 - 320605 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067502.6 |
NP_499903.3 |
8 - 9 |
1710 |
66 |
GenBank mRNA |
AY428645.1 |
- |
8 - 9 |
1744 |
66 |
View Full Product Details