Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307363.3 | NP_001294292.1 | 16 - 17 | 1771 | 103 |
NM_001307364.3 | NP_001294293.1 | 15 - 16 | 1777 | 103 | |
NM_001307365.3 | NP_001294294.1 | 14 - 15 | 1654 | 103 | |
NM_068737.2 | NP_501138.1 | 16 - 17 | 1774 | 103 | |
GenBank mRNA | U76402.1 | - | 16 - 17 | 1774 | 103 |