Assay Design
Gene Transcripts
- Gene Symbol
- sec-10
- Entrez Gene ID
-
177562
- Gene Name
- Exocyst complex component 5
- Gene Aliases
-
CELE_C33H5.9
- Chromosome Location
-
Chr.IV: 7777930 - 7782499 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068882.7 |
NP_501283.2 |
3 - 4 |
1520 |
107 |
View Full Product Details