Assay Design
Gene Transcripts
- Gene Symbol
- bbs-2
- Entrez Gene ID
-
259580
- Gene Name
- Bardet-Biedl syndrome 2 protein homolog
- Gene Aliases
-
CELE_F20D12.3
- Chromosome Location
-
Chr.IV: 7934033 - 7937749 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068924.3 |
NP_501325.2 |
4 - 5 |
1341 |
142 |
View Full Product Details