Assay Design
Gene Transcripts
- Gene Symbol
- rig-4
- Entrez Gene ID
-
177597
- Gene Name
- Protein sidekick homolog
- Gene Aliases
-
CELE_Y42H9B.2
- Chromosome Location
-
Chr.IV: 7991410 - 8004136 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068938.8 |
NP_501339.2 |
9 - 10 |
4261 |
82 |
View Full Product Details