Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038305.3 | NP_001033394.1 | 7 - 8 | 1583 | 88 |
NM_001038306.5 | NP_001033395.1 | 10 - 11 | 1961 | 88 | |
NM_001038307.5 | NP_001033396.1 | 10 - 11 | 1971 | 88 | |
NM_001038308.4 | NP_001033397.1 | 7 - 8 | 1346 | 88 | |
GenBank mRNA | AB206669.1 | - | 10 - 11 | 1898 | 88 |
AB206670.1 | - | 7 - 8 | 1346 | 88 |