Assay Design



Gene Transcripts

Gene Symbol
clec-51
Entrez Gene ID
181854
Gene Name
protein-coding
Gene Aliases
CELE_B0218.6
Chromosome Location
Chr.IV: 8161953 - 8163800 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380354.1 NP_001368439.1 2 - 3 177 90
NM_068968.7 NP_501369.1 3 - 4 573 90

View Full Product Details