Assay Design



Gene Transcripts

Gene Symbol
vit-6
Entrez Gene ID
177619
Gene Name
protein-coding
Gene Aliases
CELE_K07H8.6
Chromosome Location
Chr.IV: 8261134 - 8266459 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028103.3 NP_001023274.1 2 - 3 599 79
NM_001028105.6 NP_001023276.1 2 - 3 577 79

View Full Product Details