Assay Design



Gene Transcripts

Gene Symbol
sqv-1
Entrez Gene ID
177631
Gene Name
protein-coding
Gene Aliases
CELE_D2096.4
Chromosome Location
Chr.IV: 8361117 - 8363426 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380358.1 NP_001368459.1 2 - 3 199 83
NM_069017.8 NP_501418.1 3 - 4 370 83
GenBank mRNA AY147933.1 - 3 - 4 400 83

View Full Product Details