Assay Design



Gene Transcripts

Gene Symbol
tag-77
Entrez Gene ID
177656
Gene Name
protein-coding
Gene Aliases
CELE_C28C12.10
Chromosome Location
Chr.IV: 8503492 - 8510548 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307116.3 NP_001294045.1 9 - 10 2437 65
NM_001380369.1 NP_001368458.1 9 - 10 2239 65
NM_069054.6 NP_501455.2 11 - 12 2790 65

View Full Product Details