Assay Design



Gene Transcripts

Gene Symbol
tag-77
Entrez Gene ID
177656
Gene Name
protein-coding
Gene Aliases
CELE_C28C12.10
Chromosome Location
Chr.IV: 8503492 - 8510548 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307115.4 NP_001294044.2 1 - 2 149 92
NM_069054.6 NP_501455.2 1 - 2 150 92

View Full Product Details