Assay Design
Gene Transcripts
- Gene Symbol
- oma-1
- Entrez Gene ID
-
177703
- Gene Name
- CCCH-type zinc finger protein oma-1
- Gene Aliases
-
CELE_C09G9.6
- Chromosome Location
-
Chr.IV: 8888976 - 8890999 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069141.4 |
NP_501542.1 |
3 - 4 |
551 |
102 |
View Full Product Details