Assay Design
Gene Transcripts
- Gene Symbol
- rab-28
- Entrez Gene ID
-
189429
- Gene Name
- Ras-related protein Rab-28
- Gene Aliases
-
CELE_Y11D7A.4
- Chromosome Location
-
Chr.IV: 9242074 - 9244012 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069208.3 |
NP_501609.1 |
3 - 4 |
276 |
65 |
View Full Product Details