Assay Design



Gene Transcripts

Gene Symbol
cng-3
Entrez Gene ID
3565463
Gene Name
Cyclic nucleotide-binding domain-containing protein
Gene Aliases
CELE_F38E11.12
Chromosome Location
Chr.IV: 9475836 - 9481037 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028023.3 NP_001023194.1 2 - 3 434 128

View Full Product Details