Assay Design



Gene Transcripts

Gene Symbol
elo-7
Entrez Gene ID
186426
Gene Name
Elongation of very long chain fatty acids protein
Gene Aliases
CELE_F56H11.3
Chromosome Location
Chr.IV: 9529947 - 9531400 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268468.2 NP_001255397.1 4 - 5 685 88

View Full Product Details