Assay Design



Gene Transcripts

Gene Symbol
elt-1
Entrez Gene ID
177794
Gene Name
protein-coding
Gene Aliases
CELE_W09C2.1
Chromosome Location
Chr.IV: 9614995 - 9620280 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038346.6 NP_001033435.1 4 - 5 591 121
NM_001038347.3 NP_001033436.1 2 - 3 268 121
NM_001268473.4 NP_001255402.1 4 - 5 571 121
NM_001380414.1 NP_001368126.1 3 - 4 472 121
NM_001380415.1 NP_001368482.1 4 - 5 553 121

View Full Product Details