Assay Design



Gene Transcripts

Gene Symbol
dur-1
Entrez Gene ID
177846
Gene Name
protein-coding
Gene Aliases
CELE_F25H8.5
Chromosome Location
Chr.IV: 9921938 - 9928230 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027974.4 NP_001023145.1 13 - 14 840 69
NM_001268504.3 NP_001255433.1 16 - 17 1035 69
NM_001268505.3 NP_001255434.1 17 - 18 1101 69
NM_001268506.3 NP_001255435.1 17 - 18 1104 69
NM_001268507.3 NP_001255436.1 19 - 20 1938 69
NM_001268510.3 NP_001255439.1 10 - 11 618 69
NM_001268514.3 NP_001255443.1 6 - 7 375 69
NM_001307013.3 NP_001293942.1 13 - 14 771 69
NM_001383174.1 11 - 12 742 69
NM_001383175.1 13 - 14 840 69
NM_069385.6 NP_501786.2 17 - 18 1816 69
NM_069386.8 NP_501787.3 18 - 19 1869 69

View Full Product Details