Assay Design
Gene Transcripts
- Gene Symbol
- gst-2
- Entrez Gene ID
-
177884
- Gene Name
- Glutathione S-transferase 2
- Gene Aliases
-
CELE_K08F4.6
- Chromosome Location
-
Chr.IV: 10140330 - 10142061 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069446.4 |
NP_501847.1 |
3 - 4 |
406 |
147 |
GenBank mRNA |
AF010240.1 |
- |
2 - 3 |
348 |
147 |
View Full Product Details