Assay Design



Gene Transcripts

Gene Symbol
gon-4
Entrez Gene ID
177899
Gene Name
protein-coding
Gene Aliases
CELE_K04D7.5
Chromosome Location
Chr.IV: 10197816 - 10202993 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129314.4 NP_001122786.1 3 - 4 458 185
NM_069463.6 NP_501864.2 3 - 4 462 185

View Full Product Details