Assay Design



Gene Transcripts

Gene Symbol
abt-1
Entrez Gene ID
182849
Gene Name
protein-coding
Gene Aliases
CELE_C24F3.5
Chromosome Location
Chr.IV: 10206189 - 10214911 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306925.2 NP_001293854.1 26 - 27 4164 76
NM_001306926.3 NP_001293855.1 23 - 24 3927 76

View Full Product Details