Assay Design
Gene Transcripts
- Gene Symbol
- nas-12
- Entrez Gene ID
-
182848
- Gene Name
- Zinc metalloproteinase nas-12
- Gene Aliases
-
CELE_C24F3.3
- Chromosome Location
-
Chr.IV: 10224023 - 10226387 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069470.4 |
NP_501871.2 |
4 - 5 |
934 |
104 |
View Full Product Details