Assay Design



Gene Transcripts

Gene Symbol
unc-43
Entrez Gene ID
177921
Gene Name
protein-coding
Gene Aliases
CELE_K11E8.1
Chromosome Location
Chr.IV: 10324254 - 10350292 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028127.6 NP_001023298.1 5 - 6 639 79
NM_001268551.3 NP_001255480.1 6 - 7 658 79
NM_001372918.2 NP_001359633.1 7 - 7 719 79
NM_001380439.2 NP_001366993.1 6 - 7 779 79
NM_001380440.2 NP_001366994.1 6 - 7 733 79
NM_001380441.2 NP_001366996.1 3 - 4 482 79
NM_001380442.1 NP_001366992.1 4 - 5 563 79
NM_001380443.1 NP_001366997.1 3 - 4 328 79
NM_001383186.1 5 - 6 639 79
GenBank mRNA AF180735.1 - 4 - 5 541 79
AF233262.1 - 3 - 4 328 79
AF233263.1 - 3 - 4 328 79
AF233264.1 - 3 - 4 328 79
AF233265.1 - 3 - 4 328 79
AF233266.1 - 3 - 4 328 79
AF233267.1 - 3 - 4 328 79
AF255956.1 - 3 - 4 328 79

View Full Product Details