Assay Design



Gene Transcripts

Gene Symbol
tax-6
Entrez Gene ID
177943
Gene Name
protein-coding
Gene Aliases
CELE_C02F4.2
Chromosome Location
Chr.IV: 10492869 - 10501600 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026121.2 NP_001021292.1 3 - 4 383 74
NM_001083189.5 NP_001076658.2 3 - 4 383 74
NM_001380448.1 NP_001367052.1 2 - 3 131 74

View Full Product Details