Assay Design
Gene Transcripts
- Gene Symbol
- srd-7
- Entrez Gene ID
-
191797
- Gene Name
- Serpentine receptor class delta-7
- Gene Aliases
-
CELE_C06G8.4
- Chromosome Location
-
Chr.IV: 10785090 - 10786831 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069600.3 |
NP_502001.1 |
1 - 2 |
276 |
103 |
View Full Product Details