Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001268592.3 | NP_001255521.1 | 13 - 14 | 4012 | 105 |
NM_001268593.3 | NP_001255522.1 | 7 - 8 | 3125 | 105 | |
NM_001276852.4 | NP_001263781.1 | 11 - 12 | 3810 | 105 | |
NM_001276853.3 | NP_001263782.1 | 12 - 13 | 3928 | 105 | |
NM_001383200.1 | 13 - 14 | 4057 | 105 |