Assay Design
Gene Transcripts
- Gene Symbol
- dnc-1
- Entrez Gene ID
-
177987
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_ZK593.5
- Chromosome Location
-
Chr.IV: 10926719 - 10933934 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383200.1 |
|
4 - 5 |
637 |
76 |
View Full Product Details