Assay Design
Gene Transcripts
- Gene Symbol
- str-3
- Entrez Gene ID
-
187459
- Gene Name
- Seven TM Receptor
- Gene Aliases
-
CELE_M7.13
- Chromosome Location
-
Chr.IV: 11098448 - 11100371 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069670.2 |
NP_502071.2 |
3 - 4 |
376 |
99 |
View Full Product Details