Assay Design
Gene Transcripts
- Gene Symbol
- gtl-1
- Entrez Gene ID
-
178031
- Gene Name
- LSDAT_euk domain-containing protein
- Gene Aliases
-
CELE_C05C12.3
- Chromosome Location
-
Chr.IV: 11248884 - 11259535 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069710.6 |
NP_502111.3 |
17 - 18 |
2575 |
72 |
GenBank mRNA |
AY726541.1 |
- |
17 - 18 |
2505 |
72 |
View Full Product Details