Assay Design
Gene Transcripts
- Gene Symbol
- pfd-4
- Entrez Gene ID
-
178045
- Gene Name
- Probable prefoldin subunit 4
- Gene Aliases
-
CELE_B0035.4
- Chromosome Location
-
Chr.IV: 11315207 - 11315877 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069727.7 |
NP_502128.1 |
2 - 3 |
218 |
113 |
GenBank mRNA |
AF292051.1 |
- |
2 - 3 |
218 |
113 |
View Full Product Details