Assay Design
Gene Transcripts
- Gene Symbol
- cutl-28
- Entrez Gene ID
-
176895
- Gene Name
- CUTiclin-Like
- Gene Aliases
-
CELE_F41A4.1
- Chromosome Location
-
Chr.IV: 684497 - 688340 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067570.6 |
NP_499971.2 |
3 - 4 |
442 |
63 |
View Full Product Details