Assay Design
Gene Transcripts
- Gene Symbol
- srt-23
- Entrez Gene ID
-
190321
- Gene Name
- Serpentine Receptor, class T
- Gene Aliases
-
CELE_Y55F3C.2
- Chromosome Location
-
Chr.IV: 756721 - 759084 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067578.4 |
NP_499979.3 |
3 - 4 |
600 |
77 |
View Full Product Details