Assay Design



Gene Transcripts

Gene Symbol
madf-1
Entrez Gene ID
190316
Gene Name
protein-coding
Gene Aliases
CELE_Y55F3BR.5
Chromosome Location
Chr.IV: 830358 - 831896 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380020.2 NP_001367655.1 1 - 2 38 61

View Full Product Details