Assay Design



Gene Transcripts

Gene Symbol
lgc-33
Entrez Gene ID
176910
Gene Name
protein-coding
Gene Aliases
CELE_Y55F3BR.4
Chromosome Location
Chr.IV: 845498 - 848872 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307532.3 NP_001294461.1 6 - 7 1236 62
NM_067599.4 NP_500000.3 6 - 7 1245 62

View Full Product Details