Assay Design



Gene Transcripts

Gene Symbol
npp-20
Entrez Gene ID
176957
Gene Name
protein-coding
Gene Aliases
CELE_Y77E11A.13
Chromosome Location
Chr.IV: 1490627 - 1496974 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067685.5 NP_500086.1 4 - 5 869 77
NM_067686.5 NP_500087.1 3 - 4 253 77

View Full Product Details