Assay Design



Gene Transcripts

Gene Symbol
ced-2
Entrez Gene ID
176968
Gene Name
protein-coding
Gene Aliases
CELE_Y41D4B.13
Chromosome Location
Chr.IV: 1597318 - 1601620 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380050.1 NP_001368287.1 1 - 2 123 64
NM_067704.6 NP_500105.1 2 - 3 647 64
GenBank mRNA AF226866.1 - 2 - 3 657 64

View Full Product Details