Assay Design
Gene Transcripts
- Gene Symbol
- elt-6
- Entrez Gene ID
-
176993
- Gene Name
- GATA-type domain-containing protein
- Gene Aliases
-
CELE_F52C12.5
- Chromosome Location
-
Chr.IV: 1917977 - 1921646 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067743.7 |
NP_500144.2 |
1 - 2 |
135 |
70 |
GenBank mRNA |
AF353303.1 |
- |
1 - 2 |
135 |
70 |
View Full Product Details