Assay Design
Gene Transcripts
- Gene Symbol
- eri-5
- Entrez Gene ID
-
177029
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_Y38F2AR.1
- Chromosome Location
-
Chr.IV: 2390821 - 2396263 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067798.6 |
NP_500199.2 |
3 - 4 |
954 |
70 |
View Full Product Details