Assay Design
Gene Transcripts
- Gene Symbol
- ilys-3
- Entrez Gene ID
-
177033
- Gene Name
- Invertebrate-type lysozyme 3
- Gene Aliases
-
CELE_C45G7.3
- Chromosome Location
-
Chr.IV: 2464456 - 2465443 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067805.5 |
NP_500206.1 |
1 - 2 |
68 |
73 |
View Full Product Details