Assay Design



Gene Transcripts

Gene Symbol
lad-2
Entrez Gene ID
177078
Gene Name
protein-coding
Gene Aliases
CELE_Y54G2A.25
Chromosome Location
Chr.IV: 2964334 - 2978484 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028325.3 NP_001023496.1 16 - 17 2728 62
GenBank mRNA AF456361.1 - 16 - 17 2732 62
EF584655.1 - 16 - 17 2642 62

View Full Product Details