Assay Design



Gene Transcripts

Gene Symbol
lad-2
Entrez Gene ID
177078
Gene Name
protein-coding
Gene Aliases
CELE_Y54G2A.25
Chromosome Location
Chr.IV: 2964334 - 2978484 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028325.3 NP_001023496.1 9 - 10 1514 89
GenBank mRNA AF456361.1 - 9 - 10 1518 89
EF584655.1 - 9 - 10 1428 89

View Full Product Details