Assay Design
Gene Transcripts
- Gene Symbol
- elks-1
- Entrez Gene ID
-
177102
- Gene Name
- mammalian ELKS/CAST/ERC/Rab6 interacting protein homolog
- Gene Aliases
-
CELE_F42A6.9
- Chromosome Location
-
Chr.IV: 3342510 - 3350027 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067928.4 |
NP_500329.1 |
2 - 3 |
242 |
76 |
View Full Product Details