Assay Design
Gene Transcripts
- Gene Symbol
- vha-19
- Entrez Gene ID
-
177103
- Gene Name
- Vacuolar H ATPase
- Gene Aliases
-
CELE_Y55H10A.1
- Chromosome Location
-
Chr.IV: 3369197 - 3373485 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067931.5 |
NP_500332.1 |
1 - 2 |
85 |
66 |
View Full Product Details